4-Trifluoromethyl-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe(4-CF3)-OH, 4-(Trifluoromethyl)-L-phenylalanine

H52170-03 H52170-06 H52170-MD
AAH52170-03EA 289 USD
AAH52170-03 AAH52170-06 AAH52170-MD
4-Trifluoromethyl-L-phenylalanine 95%
4-Trifluoromethyl-L-phenylalanine

Purity: 95% CAS No.: 114926-38-4 Molecular Formula: C10H10F3NO2 Formula Weight: 233.19 Beilstein Reference: 5747857 MDL No.: MFCD00044946

Formula: C₁₀H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00044946
CAS Number: 114926-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR