3,3-Diphenyl-L-alanine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H52169-03
H52169-06
AAH52169-03EA
256
USD
AAH52169-03
AAH52169-06
AAH52169-MD
3,3-Diphenyl-L-alanine 95%
3,3-Diphenyl-L-alanine
Purity: 95%
CAS No.: 149597-92-2
Molecular Formula: C15H15NO2
Formula Weight: 241.29
UN No.: 2811
MDL No.: MFCD01631989
Formula:
C₁₅H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631989 CAS Number: 149597-92-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...