Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Thien-3-yl-L-alanine, N-FMOC protected

H52167-03 H52167-MD
AAH52167-03EA 259 USD
AAH52167-03 AAH52167-06 AAH52167-MD
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 95%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH

Purity: 95% CAS No.: 186320-06-9 Molecular Formula: C22H19NO4S Formula Weight: 393.46 Beilstein Reference: 9512976 MDL No.: MFCD00151916

Formula: C₂₂H₁₉NO₄S
MW: 393.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151916
CAS Number: 186320-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR