Boc-D-allyl-Gly-OH dicyclohexylammonium salt 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-D-allylglycine (dicyclohexylammonium) salt, 2-Allyl-N-Boc-D-glycine dicyclohexylamine salt, Boc-D-Gly(Allyl)-OH·DCHA, D-2-Allylglycine, N-BOC protected dicylohexylamine salt, Boc-D-allylglycine.DCHA, Dicyclohexylamine (R)-2-((tert-butoxycarbonyl)amino)pent-4-enoate, (R)-2-(Boc-amino)-4-pentenoic acid (dicyclohexylammonium) salt

H52127-03 H52127-06
AAH52127-03EA 81.2 USD
AAH52127-03 AAH52127-06 AAH52127-MD
Boc-D-allyl-Gly-OH dicyclohexylammonium salt 95%
Boc-D-allyl-Gly-OH dicyclohexylammonium salt

Purity: 95% CAS No.: 170899-08-8 Molecular Formula: C10H17NO4.C12H23N Formula Weight: 396.57 MDL No.: MFCD01321012

Formula: C₁₀H₁₇NO₄·C₁₂H₂₃N
MW: 396.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321012
CAS Number: 221352-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR