Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-β-HomoArg(Pbf)-OH

H52115-03 H52115-MD
AAH52115-03EA 298 USD
AAH52115-03 AAH52115-MD
Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine 95%
Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine

Purity: 95% CAS No.: 401915-53-5 Molecular Formula: C35H42N4O7S Formula Weight: 662.80 MDL No.: MFCD03788172

Formula: C₃₅H₄₂N₄O₇S
MW: 662.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788172
CAS Number: 401915-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR