3-Bromo-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-Amino-3-(3-bromophenyl)propanoic acid, H-Phe(3-Br)-OH

H52114-03 H52114-MD
AAH52114-03EA 315 USD
AAH52114-03 AAH52114-MD
3-Bromo-L-phenylalanine 95%
3-Bromo-L-phenylalanine

Purity: 95% CAS No.: 82311-69-1 Molecular Formula: C9H10BrNO2 Formula Weight: 244.09 MDL No.: MFCD06659110

Formula: C₉H₁₀BrNO₂
MW: 244.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659110
CAS Number: 82311-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR