2-Allyl-D-glycine 95%

Supplier: Thermo Scientific Chemicals

H52112-03 H52112-MD
AAH52112-03EA 225 USD
AAH52112-03 AAH52112-MD
2-Allyl-D-glycine 95%
2-Allyl-D-glycine

Purity: 95% CAS No.: 54594-06-8 Molecular Formula: C5H9NO2 Formula Weight: 115.13 MDL No.: MFCD00063104

Formula: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00063104
CAS Number: 54594-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR