(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-nitrophenyl)propanoic acid

H52101-03 H52101-06
AAH52101-03EA 281 USD
AAH52101-03 AAH52101-06 AAH52101-MD
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 95%
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

Purity: 95% CAS No.: 507472-25-5 Molecular Formula: C24H20N2O6 Formula Weight: 432.43 Melting Point: 154-156° MDL No.: MFCD03428007

Formula: C₂₄H₂₀N₂O₆
MW: 432.43 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD03428007
CAS Number: 507472-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR