L-4-Benzoylphenylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-Amino-3-(4-benzoylphenyl)propanoic acid, H-Bpa-OH, H-p-Bz-Phe-OH, H-Phe(4-Bz)-OH
H52083-03
H52083-06
H52083-MD
AAH52083-03EA
272
USD
AAH52083-03
AAH52083-06
AAH52083-MD
L-4-Benzoylphenylalanine 95%
L-4-Benzoylphenylalanine
Purity: 95%
CAS No.: 104504-45-2
Molecular Formula: C16H15NO3
Formula Weight: 269.30
MDL No.: MFCD00238102
Formula:
C₁₆H₁₅NO₃ MW: 269.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00238102 CAS Number: 104504-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...