(R)-3-(Fmoc-amino)-3-phenylpropionic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid
H52074-03
H52074-MD
AAH52074-03EA
201
USD
AAH52074-03
AAH52074-06
AAH52074-MD
(R)-3-(Fmoc-amino)-3-phenylpropionic acid 95%
(R)-3-(Fmoc-amino)-3-phenylpropionic acid
Purity: 95%
CAS No.: 220498-02-2
Molecular Formula: C24H21NO4
Formula Weight: 387.43
Beilstein Reference: 7982022
MDL No.: MFCD01311783
Formula:
C₂₄H₂₁NO₄ MW: 387.43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311783 CAS Number: 220498-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...