Fmoc-L-4-methylphe 95%
Supplier: Thermo Scientific Chemicals
H52073-03
H52073-06
AAH52073-03EA
104
USD
AAH52073-03
AAH52073-06
AAH52073-MD
Fmoc-L-4-methylphe 95%
Fmoc-L-4-methylphe
Purity: 95%
CAS No.: 199006-54-7
Molecular Formula: C25H23NO4
Formula Weight: 401.46
Beilstein Reference: 8799411
MDL No.: MFCD00270197
Formula:
C₂₅H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00270197 CAS Number: 199006-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...