(S)-3-(Boc-amino)-3-phenylpropionic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-Boc-beta3-phenylalanine, Boc-D-β-Phe-OH, (S)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid
H52072-03
H52072-06
H52072-MD
AAH52072-03EA
206
USD
AAH52072-03
AAH52072-06
AAH52072-MD
(S)-3-(Boc-amino)-3-phenylpropionic acid 95%
(S)-3-(Boc-amino)-3-phenylpropionic acid
Purity: 95%
CAS No.: 103365-47-5
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Beilstein Reference: 4695945
MDL No.: MFCD01860892
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01860892 CAS Number: 103365-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...