3,5-Difluoro-N-fmoc-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Phe(3,5-DiF)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,5-difluorophenyl)propanoic acid

H52069-03 H52069-06 H52069-MD
AAH52069-03EA 230 USD
AAH52069-03 AAH52069-06 AAH52069-MD
3,5-Difluoro-N-fmoc-L-phenylalanine 95%
3,5-Difluoro-N-fmoc-L-phenylalanine

Purity: 95% CAS No.: 205526-24-5 Molecular Formula: C24H19F2NO4 Formula Weight: 423.40 MDL No.: MFCD00797579

Formula: C₂₄H₁₉F₂NO₄
MW: 423.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797579
CAS Number: 205526-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR