Fmoc-O-t-butyl-L-β-homoserine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-β-HomoSer(tBu)-OH, Fmoc-β-HoSer(tBu)-OH
H52065-03
H52065-MD
AAH52065-03EA
386
USD
AAH52065-03
AAH52065-MD
Fmoc-O-t-butyl-L-β-homoserine 95%
Fmoc-O-t-butyl-L-beta-homoserine
Purity: 95%
CAS No.: 203854-51-7
Molecular Formula: C23H27NO5
Formula Weight: 397.47
Melting Point: 96-98°
Beilstein Reference: 8014825
MDL No.: MFCD01862865
Formula:
C₂₃H₂₇NO₅ MW: 397.48 g/mol Melting Pt: 96…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862865 CAS Number: 203854-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...