Boc-L-3,3-diphenylalanine 95%
Supplier: Thermo Scientific Chemicals
H52063-03
H52063-06
AAH52063-03EA
120
USD
AAH52063-03
AAH52063-06
AAH52063-MD
Boc-L-3,3-diphenylalanine 95%
Boc-L-3,3-diphenylalanine
Purity: 95%
CAS No.: 138662-63-2
Molecular Formula: C20H23NO4
Formula Weight: 341.41
Melting Point: 160° dec.
Beilstein Reference: 4880641
MDL No.: MFCD00191186
Formula:
C₂₀H₂₃NO₄ Melting Pt: 160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00191186 CAS Number: 138662-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...