Fmoc-L-β-homoproline 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-β-HomoPro-OH, Fmoc-β-HoPro-OH

H52060-03 H52060-MD
AAH52060-03EA 334 USD
AAH52060-03 AAH52060-MD
Fmoc-L-β-homoproline 95%
Fmoc-L-beta-homoproline

Purity: 95% CAS No.: 193693-60-6 Molecular Formula: C21H21NO4 Formula Weight: 351.40 Beilstein Reference: 7798553 MDL No.: MFCD01863058

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863058
CAS Number: 193693-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR