N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-pentafluoro-L-phenylalanine, Fmoc-Phe(F5)-OH

H52059-03 H52059-06 H52059-MD
AAH52059-03EA 151 USD
AAH52059-03 AAH52059-06 AAH52059-MD
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine

Purity: 95% CAS No.: 205526-32-5 Molecular Formula: C24H16F5NO4 Formula Weight: 477.39 MDL No.: MFCD00270612

Formula: C₂₄H₁₆F₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00270612
CAS Number: 205526-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR