Boc-3-nitro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-L-3-Nitrophenylalanine, Boc-Phe(3-NO₂)-OH, 3-Nitro-L-phenylalanine, N-BOC protected

H52058-03 H52058-06
AAH52058-03EA 144 USD
AAH52058-03 AAH52058-06 AAH52058-MD
Boc-3-nitro-L-phenylalanine 95%
Boc-3-nitro-L-phenylalanine

Purity: 95% CAS No.: 131980-29-5 Molecular Formula: C14H18N2O6 Formula Weight: 310.31 MDL No.: MFCD01317033

Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317033
CAS Number: 131980-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR