Fmoc-3-(3-pyridyl)-L-alanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-3-Pal-OH, Fmoc-β-(3-pyridyl)-Ala-OH

H52056-03 H52056-06 H52056-MD
AAH52056-03EA 171 USD
AAH52056-03 AAH52056-06 AAH52056-MD
Fmoc-3-(3-pyridyl)-L-alanine 95%
Fmoc-3-(3-pyridyl)-L-alanine

Purity: 95% CAS No.: 175453-07-3 Molecular Formula: C23H20N2O4 Formula Weight: 388.42 Beilstein Reference: 9154190 MDL No.: MFCD00144887

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144887
CAS Number: 175453-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR