Fmoc-β-(3-benzothienyl)-Ala-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-3-Ala(3-benzothienyl)-OH, Fmoc-β-(3-benzothienyl)-Ala-OH
H52054-03
H52054-06
H52054-MD
AAH52054-03EA
173
USD
AAH52054-03
AAH52054-06
AAH52054-MD
Fmoc-β-(3-benzothienyl)-Ala-OH 95%
Fmoc-beta-(3-benzothienyl)-Ala-OH
Purity: 95%
CAS No.: 177966-60-8
Molecular Formula: C26H21NO4S
Formula Weight: 443.52
Storage Temperature: 2°C to 8°C
Beilstein Reference: 9296209
MDL No.: MFCD00672562
Formula:
C₂₆H₂₁NO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00672562 CAS Number: 177966-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...