3-Methyl-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe(3-Me)-OH

H52047-03 H52047-06 H52047-MD
AAH52047-03EA 108 USD
AAH52047-03 AAH52047-06 AAH52047-MD
3-Methyl-L-phenylalanine 95%
3-Methyl-L-phenylalanine

Purity: 95% CAS No.: 114926-37-3 Molecular Formula: C10H13NO2 Formula Weight: 179.22 MDL No.: MFCD00044972

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044972
CAS Number: 114926-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR