(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-nitrophenyl)propanoic acid
H52037-03
H52037-06
H52037-MD
AAH52037-03EA
213
USD
AAH52037-03
AAH52037-06
AAH52037-MD
(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 98%
(R)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid
Purity: 98%
CAS No.: 517905-93-0
Molecular Formula: C24H20N2O6
Formula Weight: 432.43
Melting Point: 170-172°
Optical Rotation: -182°(c=1 in DMF)
MDL No.: MFCD03428045
Formula:
C₂₄H₂₀N₂O₆ MW: 432.43 g/mol Melting Pt: 170…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428045 CAS Number: 517905-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...