(R)-3-Amino-3-phenylpropionic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-D-β-Phenyl-Ala-OH, 3-Amino-3-phenylpropionic-acid, (R)-3-Amino-3-phenylpropanoic acid, (R)-b-Amino-benzenepropanoic acid
H52034-03
H52034-06
H52034-MD
AAH52034-03EA
176
USD
AAH52034-03
AAH52034-06
AAH52034-MD
(R)-3-Amino-3-phenylpropionic acid 95%
(R)-3-Amino-3-phenylpropionic acid
Purity: 95%
CAS No.: 83649-48-3
Molecular Formula: C9H11NO2
Formula Weight: 165.19
MDL No.: MFCD01076237
Formula:
C₉H₁₁NO₂ MW: 165.189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01076237 CAS Number: 13921-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...