Fmoc-4-(boc-amino)-L-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Phe(4-NHBoc)-OH, Fmoc-4-(Boc-amino)-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((tert-butoxycarbonyl)amino)phenyl)propanoic acid
H52031-03
H52031-MD
AAH52031-03EA
213
USD
AAH52031-03
AAH52031-06
AAH52031-MD
Fmoc-4-(boc-amino)-L-phenylalanine 95%
Fmoc-4-(boc-amino)-L-phenylalanine
Purity: 95%
CAS No.: 174132-31-1
Molecular Formula: C29H30N2O6
Formula Weight: 502.57
MDL No.: MFCD00237007
Formula:
C₂₉H₃₀N₂O₆ MW: 502.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00237007 CAS Number: 174132-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...