(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-3-Amino-3-(3-nitrophenyl)propanoic acid

H52027-03 H52027-06
AAH52027-03EA 199 USD
AAH52027-03 AAH52027-06 AAH52027-MD
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid

Purity: 95% CAS No.: 734529-57-8 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 MDL No.: MFCD04113693

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04113693
CAS Number: 734529-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR