Boc-3-bromo-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Phe(3-Br)-OH

H52024-03 H52024-06
AAH52024-03EA 116 USD
AAH52024-03 AAH52024-06 AAH52024-MD
Boc-3-bromo-L-phenylalanine 95%
Boc-3-bromo-L-phenylalanine

Purity: 95% CAS No.: 82278-73-7 Molecular Formula: C14H18BrNO4 Formula Weight: 344.20 MDL No.: MFCD01317710

Formula: C₁₄H₁₈BrNO₄
MW: 344.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317710
CAS Number: 82278-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR