3-Chloro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe(3-Cl)-OH

H52018-03 H52018-06
AAH52018-03EA 74.5 USD
AAH52018-03 AAH52018-06 AAH52018-MD
3-Chloro-L-phenylalanine 95%
3-Chloro-L-phenylalanine

Purity: 95% CAS No.: 80126-51-8 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 MDL No.: MFCD01075124

Formula: C₉H₁₀ClNO₂
MW: 199.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075124
CAS Number: 80126-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR