Boc-3-chloro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(3-Cl)-OH

H52015-03 H52015-06
AAH52015-03EA 66.4 USD
AAH52015-03 AAH52015-06 AAH52015-MD
Boc-3-chloro-L-phenylalanine 95%
Boc-3-chloro-L-phenylalanine

Purity: 95% CAS No.: 114873-03-9 Molecular Formula: C14H18ClNO4 Formula Weight: 299.75 MDL No.: MFCD00672515

Formula: C₁₄H₁₈ClNO₄
MW: 299.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672515
CAS Number: 114873-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR