Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-4-(Boc-aminomethyl)-L-phenylalanine

H52008-03 H52008-MD
AAH52008-03EA 232 USD
AAH52008-03 AAH52008-MD
Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%
Fmoc-4-(boc-aminomethyl)-L-phenylalanine

Purity: 95% CAS No.: 204715-91-3 Molecular Formula: C30H32N2O6 Formula Weight: 516.59 MDL No.: MFCD01317015

Formula: C₃₀H₃₂N₂O₆
MW: 516.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317015
CAS Number: 204715-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR