Fmoc-L-4,4'-biphenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-4-phenyl-Phe-OH, Fmoc-Bip(4,4')-OH

H52002-03 H52002-06 H52002-MD
AAH52002-03EA 181 USD
AAH52002-03 AAH52002-06 AAH52002-MD
Fmoc-L-4,4'-biphenylalanine 95%
Fmoc-L-4,4'-biphenylalanine

Purity: 95% CAS No.: 199110-64-0 Molecular Formula: C30H25NO4 Formula Weight: 463.52 MDL No.: MFCD00191198

Formula: C₃₀H₂₅NO₄
MW: 463.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191198
CAS Number: 199110-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR