Fmoc-Dab(Boc)-OH 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S)-2,4-Diaminobutanoic acid, 4-BOC, 2-FMOC protected
H51990-03
H51990-06
H51990-MD
AAH51990-03EA
105
USD
AAH51990-03
AAH51990-06
AAH51990-MD
Fmoc-Dab(Boc)-OH 96%
Fmoc-Dab(Boc)-OH
Purity: 96%
CAS No.: 125238-99-5
Molecular Formula: C24H28N2O6
Formula Weight: 440.50
Melting Point: 111-113°
Beilstein Reference: 6250328
Optical Rotation: -12° (c=1 in methanol)
MDL No.: MFCD00151941
Formula:
C₂₄H₂₈N₂O₆ MW: 440.496 g/mol Melting Pt: 111…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151941 CAS Number: 125238-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...