Boc-Dab(Fmoc)-OH 95%

Supplier: Thermo Scientific Chemicals

H51989-03 H51989-06
AAH51989-03EA 88 USD
AAH51989-03 AAH51989-06 AAH51989-MD
Boc-Dab(Fmoc)-OH 95%
Boc-Dab(Fmoc)-OH

Purity: 95% CAS No.: 117106-21-5 Molecular Formula: C24H28N2O6 Formula Weight: 440.50 Beilstein Reference: 4771583 MDL No.: MFCD00236845

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236845
CAS Number: 117106-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR