(R)-3-(Boc-amino)-3-phenylpropionic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, (R)-N-BOC-3-Amino-3-phenylpropanoic acid
H51984-03
H51984-MD
AAH51984-03EA
231
USD
AAH51984-03
AAH51984-06
AAH51984-MD
(R)-3-(Boc-amino)-3-phenylpropionic acid 95%
(R)-3-(Boc-amino)-3-phenylpropionic acid
Purity: 95%
CAS No.: 161024-80-2
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Beilstein Reference: 5404389
MDL No.: MFCD01320859
Formula:
C₁₄H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01320859 CAS Number: 161024-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...