N-Boc-4-methyl-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-p-Me-Phe-OH, Boc-Phe(4-Me)-OH

H51983-03 H51983-06
AAH51983-03EA 117 USD
AAH51983-03 AAH51983-06 AAH51983-MD
N-Boc-4-methyl-L-phenylalanine 95%
N-Boc-4-methyl-L-phenylalanine

Purity: 95% CAS No.: 80102-26-7 Molecular Formula: C15H21NO4 Formula Weight: 279.34 Melting Point: 84-88° Beilstein Reference: 5381557 Optical Rotation: +16° (c=1 in methanol) MDL No.: MFCD01317027

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD01317027
CAS Number: 80102-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR