(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic Acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid

H51981-03 H51981-06 H51981-MD
AAH51981-03EA 66.6 USD
AAH51981-03 AAH51981-06 AAH51981-MD
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic Acid 95%
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

Purity: 95% CAS No.: 86069-86-5 Molecular Formula: C21H21NO4 Formula Weight: 351.40 Melting Point: 153-158° Beilstein Reference: 4718405 MDL No.: MFCD00235898

Formula: C₂₁H₂₁NO₄
MW: 351.40 g/mol
Melting Pt: 153…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00235898
CAS Number: 86069-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR