(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Amino-N-Boc-L-phenylalanine, Boc-p-amino-Phe-OH, Boc-Phe(4-NH2)-OH
H51980-03
H51980-06
AAH51980-03EA
90
USD
AAH51980-03
AAH51980-06
AAH51980-MD
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Purity: 95%
CAS No.: 55533-24-9
Molecular Formula: C14H20N2O4
Formula Weight: 280.32
Melting Point: 124-126°
MDL No.: MFCD00038139
Formula:
C₁₄H₂₀N₂O₄ Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038139 CAS Number: 55533-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...