4-Iodo-L-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Phe(4-I)-OH
H51979-03
H51979-06
AAH51979-03EA
110
USD
AAH51979-03
AAH51979-06
AAH51979-MD
4-Iodo-L-phenylalanine 95%
4-Iodo-L-phenylalanine
Purity: 95%
CAS No.: 24250-85-9
Molecular Formula: C9H10INO2
Formula Weight: 291.09
Beilstein Reference: 4411317
Optical Rotation: -6.5° (c=1 in acetic acid:water (4:1))
Formula:
C₉H₁₀INO₂ MW: 291.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002602 CAS Number: 24250-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...