Fmoc-L-Phe(4-Br)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Phe(4-Br)-OH

H51978-03 H51978-06
AAH51978-03EA 102 USD
AAH51978-03 AAH51978-06 AAH51978-MD
Fmoc-L-Phe(4-Br)-OH 95%
Fmoc-L-Phe(4-Br)-OH

Purity: 95% CAS No.: 198561-04-5 Molecular Formula: C24H20BrNO4 Formula Weight: 466.33 Melting Point: 165-168° MDL No.: MFCD00273460

Formula: C₂₄H₂₀BrNO₄
MW: 466.30 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00273460
CAS Number: 198561-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR