Fmoc-4-iodo-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH

H51977-03 H51977-06
AAH51977-03EA 108 USD
AAH51977-03 AAH51977-06 AAH51977-MD
Fmoc-4-iodo-L-phenylalanine 95%
Fmoc-4-iodo-L-phenylalanine

Purity: 95% CAS No.: 82565-68-2 Molecular Formula: C24H20INO4 Formula Weight: 513.33 Melting Point: 213-217° Optical Rotation: -10° (c=1 in DMF) MDL No.: MFCD00672340

Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00672340
CAS Number: 82565-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR