Fmoc-L-β-homoleucine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-L-β-HoLeu-OH, (S)-3-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-5-methylhexanoic acid, Fmoc-L-beta-homoleucine, Fmoc-β-HomoLeu-OH, Fmoc-beta-Homoleu-OH, [(9H-Fluoren-9-ylmethoxy)carbonyl]-L-beta-homoleucine
H51976-03
H51976-MD
AAH51976-03EA
368
USD
AAH51976-03
AAH51976-MD
Fmoc-L-β-homoleucine 95%
Fmoc-L-beta-homoleucine
Purity: 95%
CAS No.: 193887-44-4
Molecular Formula: C22H25NO4
Formula Weight: 367.44
Beilstein Reference: 7723942
MDL No.: MFCD01863059
Formula:
C₂₂H₂₅NO₄ MW: 367.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863059 CAS Number: 193887-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...