Fmoc-Tic-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
H51975-03
H51975-06
AAH51975-03EA
98.8
USD
AAH51975-03
AAH51975-06
AAH51975-MD
Fmoc-Tic-OH 95%
Fmoc-Tic-OH
Purity: 95%
CAS No.: 136030-33-6
Molecular Formula: C25H21NO4
Formula Weight: 399.45
Beilstein Reference: 4827574
Optical Rotation: +26.0° (c=1 in methanol)
MDL No.: MFCD00144368
Formula:
C₂₅H₂₁NO₄ MW: 399.446 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00144368 CAS Number: 136030-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...