Fmoc-3-(2-naphthyl)-D-alanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-D-2-Nal-OH
H51974-03
H51974-06
AAH51974-03EA
106
USD
AAH51974-03
AAH51974-06
AAH51974-MD
Fmoc-3-(2-naphthyl)-D-alanine 95%
Fmoc-3-(2-naphthyl)-D-alanine
Purity: 95%
CAS No.: 138774-94-4
Molecular Formula: C28H23NO4
Formula Weight: 437.49
Beilstein Reference: 7864489
MDL No.: MFCD00077044
Formula:
C₂₈H₂₃NO₄ MW: 437.495 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077044 CAS Number: 138774-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...