Fmoc-4-fluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine

H51972-03 H51972-06 H51972-MD
AAH51972-03EA 92.6 USD
AAH51972-03 AAH51972-06 AAH51972-MD
Fmoc-4-fluoro-L-phenylalanine 95%
Fmoc-4-fluoro-L-phenylalanine

Purity: 95% CAS No.: 169243-86-1 Molecular Formula: C24H20FNO4 Formula Weight: 405.42 Beilstein Reference: 8287377 MDL No.: MFCD00191197

Formula: C₂₄H₂₀FNO₄
MW: 405.425 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191197
CAS Number: 169243-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR