4-Amino-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe(4-NH₂)-OH, H-p-Amino-Phe-OH, H-Phe(4-NH2)-OH

H51970-03 H51970-06
AAH51970-03EA 80.3 USD
AAH51970-03 AAH51970-06 AAH51970-MD
4-Amino-L-phenylalanine 95%
4-Amino-L-phenylalanine

Purity: 95% CAS No.: 943-80-6 Molecular Formula: C9H12N2O2 Formula Weight: 180.20 MDL No.: MFCD00069927

Formula: C₉H₁₂N₂O₂
MW: 180.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069927
CAS Number: 943-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR