Boc-Dap(Fmoc)-OH 98%

Supplier: Thermo Scientific Chemicals

H51964-03 H51964-06
AAH51964-03EA 110 USD
AAH51964-03 AAH51964-06
Boc-Dap(Fmoc)-OH 98%
Boc-Dap(Fmoc)-OH

Purity: 98% CAS No.: 122235-70-5 Molecular Formula: C23H26N2O6 Formula Weight: 426.47 Beilstein Reference: 4771385 MDL No.: MFCD00235897

Formula: C₂₃H₂₆N₂O₆
MW: 426.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235897
CAS Number: 122235-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR