N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Phe(4-NO2)-OH, N-Fmoc-4-nitro-L-phenylalanine
H51962-03
H51962-06
AAH51962-03EA
48.3
USD
AAH51962-03
AAH51962-06
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine
Purity: 98%
CAS No.: 95753-55-2
Molecular Formula: C24H20N2O6
Formula Weight: 432.43
Melting Point: 213-223°
Beilstein Reference: 5178656
MDL No.: MFCD00057810
Formula:
C₂₄H₂₀N₂O₆ Melting Pt: 213…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00057810 CAS Number: 95753-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...