(R)-1-Boc-Piperidine-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(+)-N-BOC-2-Piperidinecarboxylic acid, Boc-D-Homopro-OH, Boc-D-HoPro-OH, (R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

H51961-06 H51961-14
AAH51961-06EA 116 USD
AAH51961-06 AAH51961-14
(R)-1-Boc-Piperidine-2-carboxylic acid 98%
(R)-1-Boc-piperidine-2-carboxylic acid

Purity: 98% CAS No.: 28697-17-8 Molecular Formula: C11H19NO4 Formula Weight: 229.28 Melting Point: 116-119° Beilstein Reference: 480023 Optical Rotation: +68°, (c=1 in acetic acid) MDL No.: MFCD00237380

Formula: C₁₁H₁₉NO₄
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00237380
CAS Number: 28697-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR