(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Phe(4-I)-OH, Boc-p-iodo-Phe-OH
H51960-06
H51960-14
AAH51960-06EA
115
USD
AAH51960-06
AAH51960-14
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid
Purity: 98%
CAS No.: 62129-44-6
Molecular Formula: C14H18INO4
Formula Weight: 391.21
Melting Point: 150° dec.
Beilstein Reference: 4503851
Optical Rotation: +22.5° (c=1 in ethyl acetate)
MDL No.: MFCD00037119
Formula:
C₁₄H₁₈INO₄ MW: 391.2 g/mol Melting Pt: 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00037119 CAS Number: 62129-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...