4,4'-Biphenyldiboronic acid bis(pinacol)ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

H51956-03 H51956-06
AAH51956-03EA 64.2 USD
AAH51956-03 AAH51956-06
4,4'-Biphenyldiboronic acid bis(pinacol)ester 95%
4,4'-Biphenyldiboronic acid bis(pinacol) ester

Purity: 95% CAS No.: 207611-87-8 Molecular Formula: C24H32B2O4 Formula Weight: 406.14 MDL No.: MFCD08669545

Formula: C₂₄H₃₂B₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08669545
CAS Number: 207611-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR