2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 9,9-Dioctylfluorene-2,7-diboronic acid bis(pinacol) ester
H51955-03
H51955-06
AAH51955-03EA
71.8
USD
AAH51955-03
AAH51955-06
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 95%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene
Purity: 95%
CAS No.: 196207-58-6
Molecular Formula: C41H64B2O4
Formula Weight: 642.58
MDL No.: MFCD16294554
Formula:
C₄₁H₆₄B₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD16294554 CAS Number: 196207-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...